Assay Details
| Gene Symbol: fws | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259130.2 | NP_001246059.1 | 3 - 4 | 1753 | 63 | |
| NM_001299086.1 | NP_001286015.1 | 3 - 4 | 1727 | 63 | ||
| NM_001299087.1 | NP_001286016.1 | 2 - 3 | 1776 | 63 | ||
| NM_078866.4 | NP_523590.1 | 2 - 3 | 1806 | 63 | ||
| NM_165221.3 | NP_724058.1 | 3 - 4 | 1757 | 63 | ||
| GenBank mRNA | BT003257.1 | - | 2 - 3 | 1759 | 63 | |
More Information
Set Membership: |
|

fws
Genomic Map