Assay Details
| Gene Symbol: qua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165222.2 | NP_724060.1 | 3 - 4 | 886 | 109 | |
| NM_165223.3 | NP_724061.1 | 3 - 4 | 926 | 109 | ||
| GenBank mRNA | AY121684.1 | - | 3 - 4 | 877 | 109 | |
| BT100209.1 | - | 3 - 4 | 742 | 109 | ||
| U10070.1 | - | 3 - 4 | 881 | 109 | ||
More Information
Set Membership: |
|

qua
Genomic Map