Assay Details
| Gene Symbol: qua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259137.2 | NP_001246066.1 | 4 - 5 | 1852 | 61 | |
| NM_165222.2 | NP_724060.1 | 4 - 5 | 1800 | 61 | ||
| NM_165223.3 | NP_724061.1 | 4 - 5 | 1840 | 61 | ||
| GenBank mRNA | AY121684.1 | - | 4 - 5 | 1791 | 61 | |
| BT100209.1 | - | 4 - 5 | 1656 | 61 | ||
| U10070.1 | - | 4 - 5 | 1795 | 61 | ||
More Information
Set Membership: |
|

qua
Genomic Map