Assay Details
| Gene Symbol: qua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259137.2 | NP_001246066.1 | 6 - 7 | 3046 | 112 | |
| NM_165222.2 | NP_724060.1 | 6 - 7 | 2994 | 112 | ||
| NM_165223.3 | NP_724061.1 | 6 - 7 | 3034 | 112 | ||
| GenBank mRNA | AY121684.1 | - | 6 - 7 | 2985 | 112 | |
| BT100209.1 | - | 6 - 7 | 2850 | 112 | ||
| U10070.1 | - | 6 - 7 | 2989 | 112 | ||
More Information
Set Membership: |
|

qua
Genomic Map