Assay Details
| Gene Symbol: qua | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259137.2 | NP_001246066.1 | 1 - 2 | 515 | 63 | |
| NM_165223.3 | NP_724061.1 | 1 - 2 | 515 | 63 | ||
| GenBank mRNA | BT100209.1 | - | 1 - 2 | 331 | 63 | |
More Information
Set Membership: |
|

qua
Genomic Map