Assay Details
| Gene Symbol: CadN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165224.2 | NP_724068.1 | 7 - 8 | 4043 | 64 | |
| NM_165225.2 | NP_724069.1 | 7 - 8 | 4043 | 64 | ||
| NM_165226.2 | NP_724070.1 | 7 - 8 | 4043 | 64 | ||
| NM_165227.3 | NP_724071.1 | 9 - 10 | 4199 | 64 | ||
More Information
Set Membership: |
|

CadN
Genomic Map