Assay Details
| Gene Symbol: RpL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273621.2 | NP_001260550.2 | 2 - 3 | 224 | 61 | |
| NM_079948.4 | NP_524687.1 | 2 - 3 | 293 | 61 | ||
| NM_165271.3 | NP_724149.1 | 2 - 3 | 224 | 61 | ||
| GenBank mRNA | AY071208.1 | - | 2 - 3 | 223 | 61 | |
More Information
Set Membership: |
|

RpL30
Genomic Map