Assay Details
| Gene Symbol: l(2)37Cc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169541.2 | NP_001163012.2 | 2 - 3 | 550 | 62 | |
| NM_057259.6 | NP_476607.2 | 2 - 3 | 550 | 62 | ||
| NM_165281.2 | NP_724165.1 | 1 - 2 | 534 | 62 | ||
| GenBank mRNA | AY122111.1 | - | 2 - 3 | 533 | 62 | |
More Information
Set Membership: |
|

l(2)37Cc
Genomic Map