Assay Details
| Gene Symbol: Pax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038824.2 | NP_001033913.1 | 3 - 4 | 757 | 136 | |
| NM_001259160.1 | NP_001246089.1 | 2 - 3 | 271 | 136 | ||
| NM_001259161.3 | NP_001246090.1 | 3 - 4 | 401 | 136 | ||
| NM_001273644.1 | NP_001260573.1 | 2 - 3 | 427 | 136 | ||
| NM_078877.4 | NP_523601.2 | 3 - 4 | 409 | 136 | ||
| NM_165293.3 | NP_724183.1 | 2 - 3 | 551 | 136 | ||
| NM_165294.3 | NP_724184.2 | 3 - 4 | 621 | 136 | ||
| NM_165295.2 | NP_724185.1 | 2 - 3 | 217 | 136 | ||
| GenBank mRNA | BT100235.1 | - | 3 - 4 | 379 | 136 | |
| BT120186.1 | - | 2 - 3 | 271 | 136 | ||
More Information
Set Membership: |
|

Pax
Genomic Map