Assay Details
| Gene Symbol: Pax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038824.2 | NP_001033913.1 | 4 - 5 | 927 | 75 | |
| NM_001259160.1 | NP_001246089.1 | 3 - 4 | 441 | 75 | ||
| NM_001259161.3 | NP_001246090.1 | 4 - 5 | 571 | 75 | ||
| NM_001273644.1 | NP_001260573.1 | 3 - 4 | 597 | 75 | ||
| NM_078877.4 | NP_523601.2 | 4 - 5 | 579 | 75 | ||
| NM_165293.3 | NP_724183.1 | 3 - 4 | 721 | 75 | ||
| NM_165294.3 | NP_724184.2 | 4 - 5 | 791 | 75 | ||
| NM_165295.2 | NP_724185.1 | 3 - 4 | 387 | 75 | ||
| GenBank mRNA | BT088784.1 | - | 3 - 4 | 556 | 75 | |
| BT100235.1 | - | 4 - 5 | 549 | 75 | ||
| BT100283.1 | - | 3 - 4 | 556 | 75 | ||
| BT120186.1 | - | 3 - 4 | 441 | 75 | ||
More Information
Set Membership: |
|

Pax
Genomic Map