Assay Details
Gene Symbol: sNPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_165316.2 | NP_724239.1 | 4 - 5 | 1255 | 58 | |
GenBank mRNA | BT150152.1 | - | 4 - 5 | 1255 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
