Assay Details
| Gene Symbol: La | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057666.5 | NP_477014.1 | 1 - 2 | 1062 | 66 | |
| GenBank mRNA | BT150066.1 | - | 2 - 3 | 1051 | 66 | |
| L32988.1 | - | 1 - 2 | 1034 | 66 | ||
More Information
Set Membership: |
|

La
Genomic Map