Assay Details
| Gene Symbol: RtGEF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103715.4 | NP_001097185.2 | 5 - 6 | 1709 | 73 | |
| NM_001169550.2 | NP_001163021.1 | 5 - 6 | 1709 | 73 | ||
| NM_001273693.1 | NP_001260622.1 | 5 - 6 | 1349 | 73 | ||
| NM_057826.5 | NP_477174.1 | 5 - 6 | 1709 | 73 | ||
| NM_165327.3 | NP_724258.1 | 5 - 6 | 1349 | 73 | ||
| GenBank mRNA | BT082080.1 | - | 5 - 6 | 1687 | 73 | |
More Information
Set Membership: |
|

RtGEF
Genomic Map