Assay Details
| Gene Symbol: RtGEF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103714.3 | NP_001097184.2 | 7 - 8 | 4077 | 94 | |
| NM_001103715.4 | NP_001097185.2 | 6 - 7 | 2129 | 94 | ||
| NM_001169550.2 | NP_001163021.1 | 6 - 7 | 2129 | 94 | ||
| NM_001273693.1 | NP_001260622.1 | 6 - 7 | 1769 | 94 | ||
| NM_001382006.1 | NP_001369133.1 | 7 - 8 | 4077 | 94 | ||
| NM_057826.5 | NP_477174.1 | 6 - 7 | 2129 | 94 | ||
| NM_165327.3 | NP_724258.1 | 6 - 7 | 1769 | 94 | ||
| GenBank mRNA | BT082080.1 | - | 6 - 7 | 2107 | 94 | |
More Information
Set Membership: |
|

RtGEF
Genomic Map