Assay Details
| Gene Symbol: phr6-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273703.1 | NP_001260632.1 | 1 - 2 | 791 | 54 | |
| NM_001273704.1 | NP_001260633.1 | 2 - 3 | 736 | 54 | ||
| NM_057840.5 | NP_477188.1 | 2 - 3 | 908 | 54 | ||
| NM_165334.3 | NP_724274.1 | 3 - 4 | 853 | 54 | ||
| GenBank mRNA | BT044074.1 | - | 2 - 3 | 906 | 54 | |
More Information
Set Membership: |
|

phr6-4
Genomic Map