Assay Details
| Gene Symbol: Nhe2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144375.3 | NP_001137847.1 | 6 - 7 | 1754 | 73 | |
| NM_001144376.2 | NP_001137848.1 | 6 - 7 | 1754 | 73 | ||
| NM_001144377.3 | NP_001137849.1 | 6 - 7 | 1748 | 73 | ||
| NM_001144378.2 | NP_001137850.1 | 6 - 7 | 1754 | 73 | ||
| NM_001144379.3 | NP_001137851.1 | 6 - 7 | 1754 | 73 | ||
| NM_001259187.2 | NP_001246116.1 | 6 - 7 | 1754 | 73 | ||
| NM_001259188.2 | NP_001246117.1 | 6 - 7 | 1754 | 73 | ||
| NM_001273731.1 | NP_001260660.1 | 6 - 7 | 1350 | 73 | ||
| NM_001273732.1 | NP_001260661.1 | 6 - 7 | 1748 | 73 | ||
| NM_001273733.1 | NP_001260662.1 | 6 - 7 | 1748 | 73 | ||
| NM_136244.5 | NP_610088.2 | 6 - 7 | 1754 | 73 | ||
| NM_165355.4 | NP_724311.1 | 6 - 7 | 1748 | 73 | ||
| GenBank mRNA | BT133306.1 | - | 6 - 7 | 1576 | 73 | |
| Gene Symbol: CG46307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347785.1 | NP_001334745.1 | 6 - 7 | 1748 | 73 | |
More Information
Set Membership: |
|

Nhe2
Genomic Map