Assay Details
| Gene Symbol: Hr39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057584.5 | NP_476932.1 | 7 - 8 | 2838 | 72 | |
| NM_134296.3 | NP_599123.1 | 7 - 8 | 2912 | 72 | ||
| NM_165364.2 | NP_724322.1 | 7 - 8 | 2896 | 72 | ||
| GenBank mRNA | AY069676.1 | - | 7 - 8 | 2851 | 72 | |
More Information
Set Membership: |
|

Hr39
Genomic Map