Assay Details
| Gene Symbol: Mondo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169561.2 | NP_001163032.1 | 9 - 10 | 4364 | 93 | |
| NM_001169562.2 | NP_001163033.1 | 10 - 11 | 3455 | 93 | ||
| NM_001169563.2 | NP_001163034.1 | 9 - 10 | 4364 | 93 | ||
| NM_001169564.2 | NP_001163035.1 | 10 - 11 | 3876 | 93 | ||
| NM_001169565.2 | NP_001163036.1 | 9 - 10 | 3803 | 93 | ||
| NM_001169566.2 | NP_001163037.1 | 10 - 11 | 3357 | 93 | ||
| NM_001169567.2 | NP_001163038.1 | 10 - 11 | 3359 | 93 | ||
| NM_001169568.2 | NP_001163039.1 | 10 - 11 | 3595 | 93 | ||
| NM_136261.3 | NP_610105.2 | 10 - 11 | 3803 | 93 | ||
| NM_165367.2 | NP_724326.1 | 10 - 11 | 3364 | 93 | ||
| NM_165368.1 | NP_724327.1 | 8 - 9 | 2827 | 93 | ||
| GenBank mRNA | AF264754.1 | - | 9 - 10 | 3299 | 93 | |
| AY051894.2 | - | 9 - 10 | 4282 | 93 | ||
| BT133442.1 | - | 8 - 9 | 2827 | 93 | ||
More Information
Set Membership: |
|

Mondo
Genomic Map