Assay Details
| Gene Symbol: Mondo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169561.2 | NP_001163032.1 | 10 - 11 | 4492 | 90 | |
| NM_001169562.2 | NP_001163033.1 | 11 - 12 | 3583 | 90 | ||
| NM_001169563.2 | NP_001163034.1 | 10 - 11 | 4492 | 90 | ||
| NM_001169564.2 | NP_001163035.1 | 11 - 12 | 4004 | 90 | ||
| NM_001169565.2 | NP_001163036.1 | 10 - 11 | 3931 | 90 | ||
| NM_001169566.2 | NP_001163037.1 | 11 - 12 | 3485 | 90 | ||
| NM_001169567.2 | NP_001163038.1 | 11 - 12 | 3487 | 90 | ||
| NM_001169568.2 | NP_001163039.1 | 11 - 12 | 3723 | 90 | ||
| NM_136261.3 | NP_610105.2 | 11 - 12 | 3931 | 90 | ||
| NM_165367.2 | NP_724326.1 | 11 - 12 | 3492 | 90 | ||
| NM_165368.1 | NP_724327.1 | 9 - 10 | 2955 | 90 | ||
| GenBank mRNA | AF264754.1 | - | 10 - 11 | 3427 | 90 | |
| AY051894.2 | - | 10 - 11 | 4410 | 90 | ||
| BT133442.1 | - | 9 - 10 | 2955 | 90 | ||
More Information
Set Membership: |
|

Mondo
Genomic Map