Assay Details
| Gene Symbol: Mondo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169561.2 | NP_001163032.1 | 7 - 8 | 3626 | 61 | |
| NM_001169562.2 | NP_001163033.1 | 8 - 9 | 2717 | 61 | ||
| NM_001169563.2 | NP_001163034.1 | 7 - 8 | 3626 | 61 | ||
| NM_001169564.2 | NP_001163035.1 | 8 - 9 | 3138 | 61 | ||
| NM_001169565.2 | NP_001163036.1 | 7 - 8 | 3065 | 61 | ||
| NM_001169566.2 | NP_001163037.1 | 8 - 9 | 2619 | 61 | ||
| NM_001169567.2 | NP_001163038.1 | 8 - 9 | 2621 | 61 | ||
| NM_001169568.2 | NP_001163039.1 | 8 - 9 | 2857 | 61 | ||
| NM_136261.3 | NP_610105.2 | 8 - 9 | 3065 | 61 | ||
| NM_165367.2 | NP_724326.1 | 8 - 9 | 2626 | 61 | ||
| NM_165368.1 | NP_724327.1 | 6 - 7 | 2089 | 61 | ||
| GenBank mRNA | AF264754.1 | - | 7 - 8 | 2561 | 61 | |
| AY051894.2 | - | 7 - 8 | 3544 | 61 | ||
| BT133442.1 | - | 6 - 7 | 2089 | 61 | ||
More Information
Set Membership: |
|

Mondo
Genomic Map