Assay Details
| Gene Symbol: crc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273743.1 | NP_001260672.1 | 5 - 6 | 1199 | 66 | |
| NM_001273744.1 | NP_001260673.1 | 4 - 5 | 833 | 66 | ||
| NM_080158.3 | NP_524897.1 | 4 - 5 | 860 | 66 | ||
| NM_165374.2 | NP_724333.1 | 2 - 3 | 1470 | 66 | ||
| GenBank mRNA | AY069092.1 | - | 2 - 3 | 1172 | 66 | |
| BT099797.1 | - | 4 - 5 | 857 | 66 | ||
| Gene Symbol: CG46314 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347792.1 | NP_001334746.1 | 2 - 3 | 1470 | 66 | |
More Information
Set Membership: |
|

crc
Genomic Map