Assay Details
| Gene Symbol: Src42A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057501.4 | NP_476849.1 | 9 - 10 | 1923 | 65 | |
| NM_136347.2 | NP_610191.2 | 9 - 10 | 5009 | 65 | ||
| GenBank mRNA | AY058652.1 | - | 9 - 10 | 1887 | 65 | |
| BT044565.1 | - | 10 - 11 | 1791 | 65 | ||
More Information
Set Membership: |
|

Src42A
Genomic Map