Assay Details
| Gene Symbol: Src42A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057501.4 | NP_476849.1 | 3 - 4 | 1086 | 74 | |
| NM_136347.2 | NP_610191.2 | 3 - 4 | 4172 | 74 | ||
| GenBank mRNA | AY058652.1 | - | 3 - 4 | 1050 | 74 | |
| BT044565.1 | - | 4 - 5 | 954 | 74 | ||
More Information
Set Membership: |
|

Src42A
Genomic Map