Assay Details
| Gene Symbol: Pngl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136348.3 | NP_610192.1 | 2 - 3 | 561 | 80 | |
| NM_165454.3 | NP_724444.3 | 2 - 3 | 561 | 80 | ||
| GenBank mRNA | AF250926.1 | - | 2 - 3 | 529 | 80 | |
More Information
Set Membership: |
|

Pngl
Genomic Map