Assay Details
| Gene Symbol: Pld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144138.2 | NP_001137610.1 | 5 - 6 | 2286 | 70 | |
| NM_078903.4 | NP_523627.3 | 4 - 5 | 1965 | 70 | ||
| NM_165469.4 | NP_724469.2 | 5 - 6 | 2127 | 70 | ||
| NM_165470.3 | NP_724470.2 | 4 - 5 | 2615 | 70 | ||
| NM_165471.3 | NP_724471.2 | 4 - 5 | 2339 | 70 | ||
| NM_165472.3 | NP_724472.3 | 4 - 5 | 1911 | 70 | ||
| GenBank mRNA | AF145640.2 | - | 4 - 5 | 2577 | 70 | |
| BT046138.1 | - | 4 - 5 | 1993 | 70 | ||
More Information
Set Membership: |
|

Pld
Genomic Map