Assay Details
| Gene Symbol: Opbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165478.3 | NP_724479.1 | 4 - 5 | 1735 | 101 | |
| GenBank mRNA | BT022170.1 | - | 4 - 5 | 1689 | 101 | |
More Information
Set Membership: |
|

Opbp
Genomic Map