Assay Details
| Gene Symbol: Rpp25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259221.2 | NP_001246150.1 | 4 - 5 | 1011 | 73 | |
| NM_136383.4 | NP_610227.2 | 4 - 5 | 517 | 73 | ||
| NM_165484.3 | NP_724490.1 | 4 - 5 | 517 | 73 | ||
| GenBank mRNA | BT044425.1 | - | 4 - 5 | 484 | 73 | |
More Information
Set Membership: |
|

Rpp25
Genomic Map