Assay Details
| Gene Symbol: Epac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103732.2 | NP_001097202.2 | 13 - 14 | 2101 | 63 | |
| NM_001273807.1 | NP_001260736.1 | 10 - 11 | 1745 | 63 | ||
| NM_001273808.1 | NP_001260737.2 | 9 - 10 | 1364 | 63 | ||
| NM_165490.2 | NP_724498.1 | 10 - 11 | 1746 | 63 | ||
| GenBank mRNA | AY058255.1 | - | 10 - 11 | 1729 | 63 | |
More Information
Set Membership: |
|

Epac
Genomic Map