Assay Details
| Gene Symbol: Epac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103732.2 | NP_001097202.2 | 14 - 15 | 2270 | 67 | |
| NM_001273807.1 | NP_001260736.1 | 11 - 12 | 1914 | 67 | ||
| NM_001273808.1 | NP_001260737.2 | 10 - 11 | 1533 | 67 | ||
| NM_165490.2 | NP_724498.1 | 11 - 12 | 1915 | 67 | ||
| GenBank mRNA | AY058255.1 | - | 11 - 12 | 1898 | 67 | |
More Information
Set Membership: |
|

Epac
Genomic Map