Assay Details
| Gene Symbol: Epac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103732.2 | NP_001097202.2 | 7 - 8 | 1215 | 51 | |
| NM_001273807.1 | NP_001260736.1 | 4 - 5 | 859 | 51 | ||
| NM_001273808.1 | NP_001260737.2 | 3 - 4 | 478 | 51 | ||
| NM_165490.2 | NP_724498.1 | 4 - 5 | 860 | 51 | ||
| GenBank mRNA | AY058255.1 | - | 4 - 5 | 843 | 51 | |
More Information
Set Membership: |
|

Epac
Genomic Map