Assay Details
| Gene Symbol: Epac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103732.2 | NP_001097202.2 | 9 - 10 | 1442 | 60 | |
| NM_001273807.1 | NP_001260736.1 | 6 - 7 | 1086 | 60 | ||
| NM_001273808.1 | NP_001260737.2 | 5 - 6 | 705 | 60 | ||
| NM_165490.2 | NP_724498.1 | 6 - 7 | 1087 | 60 | ||
| GenBank mRNA | AY058255.1 | - | 6 - 7 | 1070 | 60 | |
More Information
Set Membership: |
|

Epac
Genomic Map