Assay Details
| Gene Symbol: Epac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103732.2 | NP_001097202.2 | 11 - 12 | 1786 | 95 | |
| NM_001273807.1 | NP_001260736.1 | 8 - 9 | 1430 | 95 | ||
| NM_001273808.1 | NP_001260737.2 | 7 - 8 | 1049 | 95 | ||
| NM_165490.2 | NP_724498.1 | 8 - 9 | 1431 | 95 | ||
| GenBank mRNA | AY058255.1 | - | 8 - 9 | 1414 | 95 | |
More Information
Set Membership: |
|

Epac
Genomic Map