Assay Details
| Gene Symbol: Epac | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103732.2 | NP_001097202.2 | 12 - 13 | 1929 | 67 | |
| NM_001273807.1 | NP_001260736.1 | 9 - 10 | 1573 | 67 | ||
| NM_001273808.1 | NP_001260737.2 | 8 - 9 | 1192 | 67 | ||
| NM_165490.2 | NP_724498.1 | 9 - 10 | 1574 | 67 | ||
| GenBank mRNA | AY058255.1 | - | 9 - 10 | 1557 | 67 | |
More Information
Set Membership: |
|

Epac
Genomic Map