Assay Details
| Gene Symbol: pk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165508.2 | NP_724534.1 | 2 - 3 | 1418 | 84 | |
| NM_165509.2 | NP_724535.1 | 2 - 3 | 1286 | 84 | ||
| NM_165512.2 | NP_724538.1 | 3 - 4 | 2401 | 84 | ||
More Information
Set Membership: |
|

pk
Genomic Map