Assay Details
| Gene Symbol: didum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057838.4 | NP_477186.1 | 12 - 13 | 4170 | 87 | |
| NM_165535.2 | NP_724569.1 | 13 - 14 | 4194 | 87 | ||
| NM_165536.3 | NP_724570.1 | 11 - 12 | 4072 | 87 | ||
More Information
Set Membership: |
|

didum
Genomic Map