Assay Details
| Gene Symbol: didum | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057838.4 | NP_477186.1 | 14 - 15 | 4815 | 66 | |
| NM_165535.2 | NP_724569.1 | 15 - 16 | 4839 | 66 | ||
| NM_165536.3 | NP_724570.1 | 13 - 14 | 4717 | 66 | ||
More Information
Set Membership: |
|

didum
Genomic Map