Assay Details
| Gene Symbol: Gdap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259262.2 | NP_001246191.1 | 4 - 5 | 523 | 65 | |
| NM_001273860.2 | NP_001260789.1 | 2 - 3 | 780 | 65 | ||
| NM_001273861.2 | NP_001260790.1 | 10 - 11 | 1698 | 65 | ||
| NM_165552.5 | NP_724597.1 | 10 - 11 | 1493 | 65 | ||
| NM_165553.5 | NP_724598.1 | 4 - 5 | 527 | 65 | ||
| NM_165554.4 | NP_724599.1 | 2 - 3 | 780 | 65 | ||
| GenBank mRNA | AY122088.1 | - | 10 - 11 | 1479 | 65 | |
| BT120068.1 | - | 4 - 5 | 500 | 65 | ||
| BT133240.1 | - | 2 - 3 | 778 | 65 | ||
More Information
Set Membership: |
|

Gdap2
Genomic Map