Assay Details
| Gene Symbol: sax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259264.2 | NP_001246193.1 | 6 - 7 | 1419 | 75 | |
| NM_078928.4 | NP_523652.2 | 5 - 6 | 1383 | 75 | ||
| NM_165559.2 | NP_724606.1 | 5 - 6 | 1684 | 75 | ||
| GenBank mRNA | AY058363.1 | - | 5 - 6 | 1684 | 75 | |
| L33790.1 | - | 5 - 6 | 1381 | 75 | ||
More Information
Set Membership: |
|

sax
Genomic Map