Assay Details
| Gene Symbol: Pabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057554.4 | NP_476902.1 | 3 - 4 | 908 | 58 | |
| NM_165589.3 | NP_724648.1 | 3 - 4 | 908 | 58 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabp2| Gene Symbol: Pabp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057554.4 | NP_476902.1 | 3 - 4 | 908 | 58 | |
| NM_165589.3 | NP_724648.1 | 3 - 4 | 908 | 58 | ||
Set Membership: |
|