Assay Details
| Gene Symbol: UQCR-11L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273882.1 | NP_001260811.1 | 1 | 268 | 72 | |
| NM_165616.3 | NP_724697.1 | 2 | 212 | 72 | ||
| GenBank mRNA | BT023630.1 | - | 2 | 206 | 72 | |
| KX531341.1 | - | 1 | 152 | 72 | ||
More Information
Set Membership: |
|

UQCR-11L
Genomic Map