Assay Details
| Gene Symbol: Sec31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299285.1 | NP_001286214.1 | 11 - 12 | 3737 | 128 | |
| NM_001299286.1 | NP_001286215.1 | 11 - 12 | 3740 | 128 | ||
| NM_136570.4 | NP_610414.1 | 10 - 11 | 3680 | 128 | ||
| NM_165630.3 | NP_724716.1 | 10 - 11 | 3683 | 128 | ||
More Information
Set Membership: |
|

Sec31
Genomic Map