Assay Details
| Gene Symbol: Pgi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078939.3 | NP_523663.1 | 1 - 2 | 638 | 71 | |
| NM_165635.2 | NP_724723.1 | 2 - 3 | 499 | 71 | ||
| NM_165636.2 | NP_724724.1 | 1 - 2 | 455 | 71 | ||
| GenBank mRNA | AY051468.1 | - | 1 - 2 | 629 | 71 | |
More Information
Set Membership: |
|

Pgi
Genomic Map