Assay Details
| Gene Symbol: CG30344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165655.3 | NP_724759.1 | 5 - 6 | 1171 | 67 | |
| GenBank mRNA | AY051590.1 | - | 5 - 6 | 1165 | 67 | |
More Information
Set Membership: |
|

CG30344
Genomic Map