Assay Details
| Gene Symbol: CG30344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_165655.3 | NP_724759.1 | 8 - 9 | 1788 | 71 | |
| GenBank mRNA | AY051590.1 | - | 8 - 9 | 1782 | 71 | |
More Information
Set Membership: |
|

CG30344
Genomic Map