Assay Details
| Gene Symbol: VhaAC45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136626.5 | NP_610470.1 | 4 - 5 | 570 | 97 | |
| NM_165665.2 | NP_724770.1 | 3 - 4 | 658 | 97 | ||
| GenBank mRNA | BT003279.1 | - | 3 - 4 | 628 | 97 | |
More Information
Set Membership: |
|

VhaAC45
Genomic Map