Assay Details
| Gene Symbol: Not1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103772.3 | NP_001097242.1 | 9 - 10 | 8128 | 64 | |
| NM_001103773.3 | NP_001097243.1 | 9 - 10 | 8122 | 64 | ||
| NM_001103774.3 | NP_001097244.1 | 9 - 10 | 7629 | 64 | ||
| NM_001299323.1 | NP_001286252.1 | 8 - 9 | 7619 | 64 | ||
| NM_165678.4 | NP_724798.3 | 9 - 10 | 8128 | 64 | ||
More Information
Set Membership: |
|

Not1
Genomic Map