Assay Details
| Gene Symbol: Smyd4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136656.3 | NP_610500.2 | 1 - 2 | 707 | 87 | |
| NM_165681.2 | NP_724802.1 | 2 - 3 | 748 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smyd4-1| Gene Symbol: Smyd4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136656.3 | NP_610500.2 | 1 - 2 | 707 | 87 | |
| NM_165681.2 | NP_724802.1 | 2 - 3 | 748 | 87 | ||
Set Membership: |
|