Assay Details
| Gene Symbol: Smyd4-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_136656.3 | NP_610500.2 | 3 - 4 | 2046 | 82 | |
| NM_165681.2 | NP_724802.1 | 4 - 5 | 2087 | 82 | ||
More Information
Set Membership: |
|

Smyd4-1
Genomic Map