Assay Details
| Gene Symbol: PCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169632.2 | NP_001163103.1 | 3 - 4 | 444 | 78 | |
| NM_136683.3 | NP_610527.1 | 2 - 3 | 219 | 78 | ||
| NM_165705.3 | NP_724837.1 | 2 - 3 | 516 | 78 | ||
| NM_165706.2 | NP_724838.1 | 2 - 3 | 261 | 78 | ||
| NM_165707.3 | NP_724839.1 | 3 - 4 | 394 | 78 | ||
| NM_165708.2 | NP_724840.1 | 1 - 2 | 326 | 78 | ||
| NM_165709.2 | NP_724841.1 | 2 - 3 | 219 | 78 | ||
| NM_165710.3 | NP_724842.1 | 2 - 3 | 516 | 78 | ||
| NM_165711.2 | NP_724843.1 | 2 - 3 | 261 | 78 | ||
| NM_165712.2 | NP_724844.1 | 1 - 2 | 326 | 78 | ||
| GenBank mRNA | AF132152.1 | - | 2 - 3 | 470 | 78 | |
More Information
Set Membership: |
|

PCB
Genomic Map