Assay Details
| Gene Symbol: PCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169632.2 | NP_001163103.1 | 7 - 8 | 3691 | 63 | |
| NM_136683.3 | NP_610527.1 | 5 - 6 | 3418 | 63 | ||
| NM_165705.3 | NP_724837.1 | 5 - 6 | 3715 | 63 | ||
| NM_165706.2 | NP_724838.1 | 5 - 6 | 3460 | 63 | ||
| NM_165707.3 | NP_724839.1 | 6 - 7 | 3593 | 63 | ||
| NM_165708.2 | NP_724840.1 | 4 - 5 | 3525 | 63 | ||
| NM_165709.2 | NP_724841.1 | 6 - 7 | 3466 | 63 | ||
| NM_165710.3 | NP_724842.1 | 6 - 7 | 3763 | 63 | ||
| NM_165711.2 | NP_724843.1 | 6 - 7 | 3508 | 63 | ||
| NM_165712.2 | NP_724844.1 | 5 - 6 | 3573 | 63 | ||
| GenBank mRNA | AF132152.1 | - | 5 - 6 | 3669 | 63 | |
More Information
Set Membership: |
|

PCB
Genomic Map